Assay Details
| Gene Symbol: CENPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087306.4 | XP_001087306.1 | 866 | 79 | ||
| GenBank mRNA | JU478917.1 | - | 177 | 79 | ||
| JV049839.1 | - | 85 | 79 | |||
More Information
Set Membership: |
|

CENPA
Genomic Map