Assay Details
| Gene Symbol: SLC31A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117051.2 | XP_014972537.1 | 1199 | 105 | ||
| XM_015117052.2 | XP_014972538.1 | 580 | 105 | |||
| XM_028834968.1 | XP_028690801.1 | 2302 | 105 | |||
| GenBank mRNA | JU322897.1 | - | 105 | |||
| JU472705.1 | - | 105 | ||||
More Information
Set Membership: |
|

SLC31A2
Genomic Map