Assay Details
| Gene Symbol: CRYBA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166011.3 | NP_001159483.1 | 333 | 85 | ||
| XM_028827383.1 | XP_028683216.1 | 990 | 85 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CRYBA4
Genomic Map