Assay Details
| Gene Symbol: LOC716161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112945.4 | XP_001112945.2 | 853 | 95 | ||
| XM_028827512.1 | XP_028683345.1 | 853 | 95 | |||
| GenBank mRNA | JU481815.1 | - | 256 | 95 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC716161
Genomic Map