Assay Details
| Gene Symbol: LOC712405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116399.2 | XP_014971885.2 | 408 | 70 | ||
| GenBank mRNA | JU482548.1 | - | 53 | 70 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC712405
Genomic Map