Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133775.2 | XP_014989261.1 | 968 | 179 | ||
| XM_015133776.2 | XP_014989262.1 | 1140 | 179 | |||
| GenBank mRNA | JU333933.1 | - | 809 | 179 | ||
| JU475116.1 | - | 803 | 179 | |||
| JV046725.1 | - | 869 | 179 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GARS1
Genomic Map