Assay Details
| Gene Symbol: LOC694538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085311.4 | XP_001085311.2 | 966 | 88 | ||
| GenBank mRNA | JU475138.1 | - | 187 | 88 | ||
| JU475139.1 | - | 187 | 88 | |||
| JU475140.1 | - | 187 | 88 | |||
| JU475143.1 | - | 187 | 88 | |||
| JU484358.1 | - | 187 | 88 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_001085311.4 transcripts.
This assay is in the 5'UTR region of the XM_001085311.4 transcripts.
Set Membership: |
|

LOC694538
Genomic Map