Assay Details
| Gene Symbol: GNA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266856.1 | NP_001253785.1 | 350 | 68 | ||
| GenBank mRNA | JU326996.1 | - | 253 | 68 | ||
| JU475225.1 | - | 350 | 68 | |||
| JV578755.1 | - | 393 | 68 | |||
More Information
Set Membership: |
|

GNA11
Genomic Map