Assay Details
| Gene Symbol: HSPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106968.4 | XP_001106968.1 | 1260 | 80 | ||
| XM_015140776.2 | XP_014996262.1 | 1250 | 80 | |||
| GenBank mRNA | JU334298.1 | - | 1206 | 80 | ||
| JU472316.1 | - | 1260 | 80 | |||
| JV044802.1 | - | 1231 | 80 | |||
| JV601059.1 | - | 1256 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HSPA4
Genomic Map