Assay Details
| Gene Symbol: ITPKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115115.2 | XP_014970601.2 | 2797 | 81 | ||
| GenBank mRNA | JU343091.1 | - | 1935 | 81 | ||
| JU475499.1 | - | 2581 | 81 | |||
| JV505728.1 | - | 939 | 81 | |||
More Information
Set Membership: |
|

ITPKB
Genomic Map