Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266363.1 | NP_001253292.1 | 465 | 101 | ||
| XM_015143706.2 | XP_014999192.1 | 597 | 101 | |||
| GenBank mRNA | JU334469.1 | - | 512 | 101 | ||
| JU475566.1 | - | 465 | 101 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LGALS3
Genomic Map