Assay Details
| Gene Symbol: LOC114674161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028840024.1 | XP_028695857.1 | 385 | 79 | ||
| Gene Symbol: LOC694781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839350.1 | XP_028695183.1 | 386 | 79 | ||
More Information
Set Membership: |
|

LOC114674161
Genomic Map