Assay Details
| Gene Symbol: LTBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265736.1 | NP_001252665.1 | 580 | 98 | ||
| XM_015150804.2 | XP_015006290.1 | 1121 | 98 | |||
| XM_015150805.2 | XP_015006291.1 | 1040 | 98 | |||
| GenBank mRNA | JU325342.1 | - | 308 | 98 | ||
| JV565119.1 | - | 580 | 98 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LTBR
Genomic Map