Assay Details
| Gene Symbol: CXCL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032936.1 | NP_001028108.1 | 279 | 99 | ||
| XM_015138438.2 | XP_014993924.1 | 438 | 99 | |||
| GenBank mRNA | AY044445.1 | - | 279 | 99 | ||
| JU486844.1 | - | 310 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CXCL9
Genomic Map