Assay Details
| Gene Symbol: MMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115555.2 | XP_014971041.1 | 1363 | 119 | ||
| XM_015115556.2 | XP_014971042.1 | 1294 | 119 | |||
More Information
Set Membership: |
|

MMP8
Genomic Map