Assay Details
| Gene Symbol: IRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266270.1 | NP_001253199.1 | 230 | 85 | ||
| XM_015135403.2 | XP_014990889.1 | 261 | 85 | |||
| XM_028846902.1 | XP_028702735.1 | 261 | 85 | |||
| GenBank mRNA | JU475493.1 | - | 230 | 85 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IRF4
Genomic Map