Assay Details
Gene Symbol: NPAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112801.3 | XP_001112801.1 | 724 | 125 | ||
XM_015124326.2 | XP_014979812.1 | 700 | 125 | |||
XM_015124328.2 | XP_014979814.1 | 700 | 125 | |||
XM_015124329.2 | XP_014979815.1 | 700 | 125 | |||
XM_028839339.1 | XP_028695172.1 | 700 | 125 | |||
GenBank mRNA | JV617853.1 | - | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
