Assay Details
| Gene Symbol: NPTX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266044.1 | NP_001252973.1 | 515 | 59 | ||
| GenBank mRNA | JU320283.1 | - | 515 | 59 | ||
| JU320284.1 | - | 515 | 59 | |||
| JV669194.1 | - | 596 | 59 | |||
| JV715946.1 | - | 154 | 59 | |||
More Information
Set Membership: |
|

NPTX1
Genomic Map