Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094789.4 | XP_001094789.1 | 477 | 94 | ||
| XM_001095022.4 | XP_001095022.1 | 251 | 94 | |||
| XM_015140249.2 | XP_014995735.1 | 195 | 94 | |||
| GenBank mRNA | JU475761.1 | - | 147 | 94 | ||
| JV062221.1 | - | 127 | 94 | |||
More Information
Set Membership: |
|

OCLN
Genomic Map