Assay Details
Gene Symbol: LOC114669825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU487421.1 | - | 99 | 107 |
Gene Symbol: ODF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001110960.4 | XP_001110960.2 | 128 | 107 | ||
XM_015116688.2 | XP_014972174.1 | 153 | 107 | |||
XM_015116693.2 | XP_014972179.1 | 128 | 107 | |||
XM_015116700.2 | XP_014972186.1 | 128 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001110960.4, XM_015116688.2, XM_015116693.2, XM_015116700.2 transcripts.
Set Membership: |
![]() ![]() |
