Assay Details
Gene Symbol: POU6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151732.2 | XP_015007218.1 | 2308 | 83 | ||
XM_015151734.2 | XP_015007220.1 | 2180 | 83 | |||
XM_015151735.2 | XP_015007221.1 | 1977 | 83 | |||
XM_015151736.2 | XP_015007222.1 | 1455 | 83 | |||
XM_015151737.2 | XP_015007223.2 | 1382 | 83 | |||
XM_028829490.1 | XP_028685323.1 | 2378 | 83 | |||
XM_028829491.1 | XP_028685324.1 | 2157 | 83 | |||
GenBank mRNA | JV704232.1 | - | 1036 | 83 | ||
JV704234.1 | - | 1036 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
