Assay Details
| Gene Symbol: PRKAR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134667.2 | XP_014990153.1 | 1331 | 85 | ||
| GenBank mRNA | JV047622.1 | - | 1185 | 85 | ||
| JV587085.1 | - | 1198 | 85 | |||
| JV621975.1 | - | 1235 | 85 | |||
More Information
Set Membership: |
|

PRKAR2B
Genomic Map