Assay Details
Gene Symbol: PTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122300.2 | XP_014977786.1 | 1019 | 109 | ||
XM_015122301.2 | XP_014977787.1 | 533 | 109 | |||
XM_015122302.2 | XP_014977788.1 | 1019 | 109 | |||
XM_015122303.2 | XP_014977789.1 | 533 | 109 | |||
XM_015122304.2 | XP_014977790.1 | 491 | 109 | |||
GenBank mRNA | JU335603.1 | - | 525 | 109 | ||
JU335604.1 | - | 525 | 109 | |||
JV047742.1 | - | 544 | 109 | |||
JV047743.1 | - | 544 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
