Assay Details
| Gene Symbol: RAB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266792.2 | NP_001253721.1 | 91 | 119 | ||
| GenBank mRNA | JU321715.1 | - | 34 | 119 | ||
| JV489494.1 | - | 32 | 119 | |||
| JV624558.1 | - | 177 | 119 | |||
| JV667301.1 | - | 91 | 119 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266792.2 transcripts.
Set Membership: |
|

RAB3B
Genomic Map