Assay Details
| Gene Symbol: RAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266302.1 | NP_001253231.1 | 1074 | 66 | ||
| XM_015130204.2 | XP_014985690.1 | 1095 | 66 | |||
| XM_015130206.2 | XP_014985692.1 | 983 | 66 | |||
| XM_028843088.1 | XP_028698921.1 | 1096 | 66 | |||
| XM_028843089.1 | XP_028698922.1 | 922 | 66 | |||
| XM_028843090.1 | XP_028698923.1 | 1156 | 66 | |||
| XM_028843091.1 | XP_028698924.1 | 838 | 66 | |||
| XM_028843092.1 | XP_028698925.1 | 1003 | 66 | |||
| GenBank mRNA | JU320542.1 | - | 1011 | 66 | ||
| JU476632.1 | - | 1039 | 66 | |||
| JV047817.1 | - | 1074 | 66 | |||
| JV657736.1 | - | 1089 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAF1
Genomic Map