Assay Details
| Gene Symbol: MRPL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266331.1 | NP_001253260.1 | 368 | 97 | ||
| GenBank mRNA | JU335818.1 | - | 338 | 97 | ||
| JU476698.1 | - | 361 | 97 | |||
| JV047874.1 | - | 368 | 97 | |||
| JV658160.1 | - | 378 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPL12
Genomic Map