Assay Details
| Gene Symbol: XCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032947.1 | NP_001028119.1 | 178 | 128 | ||
| GenBank mRNA | AF449285.1 | - | 178 | 128 | ||
| JV694538.1 | - | 158 | 128 | |||
| KT359585.1 | - | 213 | 128 | |||
| Gene Symbol: LOC100423131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850129.1 | XP_028705962.1 | 910 | 128 | ||
| XM_028850130.1 | XP_028705963.1 | 1155 | 128 | |||
| XM_028850131.1 | XP_028705964.1 | 1062 | 128 | |||
| XM_028850132.1 | XP_028705965.1 | 1216 | 128 | |||
| XM_028850133.1 | XP_028705966.1 | 971 | 128 | |||
| XM_028850134.1 | XP_028705967.1 | 789 | 128 | |||
| XM_028850136.1 | XP_028705969.1 | 1034 | 128 | |||
| XM_028850137.1 | XP_028705970.1 | 732 | 128 | |||
| GenBank mRNA | JV453160.1 | - | 196 | 128 | ||
| JV554243.1 | - | 192 | 128 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

XCL1
Genomic Map