Assay Details
| Gene Symbol: SHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261487.1 | NP_001248416.1 | 1783 | 142 | ||
| XM_015110149.2 | XP_014965635.1 | 4322 | 142 | |||
| XM_015110167.2 | XP_014965653.1 | 1529 | 142 | |||
| XM_028846070.1 | XP_028701903.1 | 1526 | 142 | |||
| GenBank mRNA | JU326924.1 | - | 1268 | 142 | ||
| JV047941.1 | - | 1783 | 142 | |||
| JV068724.1 | - | 1492 | 142 | |||
| JV635393.1 | - | 1760 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SHC1
Genomic Map