Assay Details
| Gene Symbol: SLC2A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146614.2 | XP_015002100.1 | 1211 | 54 | ||
| XM_028840171.1 | XP_028696004.1 | 1165 | 54 | |||
| XM_028840175.1 | XP_028696008.1 | 1210 | 54 | |||
| GenBank mRNA | JU476870.1 | - | 1124 | 54 | ||
| JV721486.1 | - | 1102 | 54 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC2A5
Genomic Map