Assay Details
| Gene Symbol: SLC16A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257704.1 | NP_001244633.1 | 473 | 106 | ||
| XM_015147143.2 | XP_015002629.2 | 398 | 106 | |||
| XM_015147153.2 | XP_015002639.2 | 1054 | 106 | |||
| GenBank mRNA | JU323021.1 | - | 473 | 106 | ||
| JU473026.1 | - | 522 | 106 | |||
| JV721619.1 | - | 481 | 106 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC16A1
Genomic Map