Assay Details
| Gene Symbol: SPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114298.2 | XP_014969784.1 | 634 | 146 | ||
| XM_015114299.2 | XP_014969785.1 | 582 | 146 | |||
| XM_028833436.1 | XP_028689269.1 | 638 | 146 | |||
| XM_028833437.1 | XP_028689270.1 | 636 | 146 | |||
| GenBank mRNA | JU320639.1 | - | 516 | 146 | ||
More Information
Set Membership: |
|

SPI1
Genomic Map