Assay Details
| Gene Symbol: SPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260549.1 | NP_001247478.1 | 599 | 71 | ||
| GenBank mRNA | JU327213.1 | - | 481 | 71 | ||
| JV045667.1 | - | 599 | 71 | |||
| JV636333.1 | - | 624 | 71 | |||
| JV704653.1 | - | 612 | 71 | |||
More Information
Set Membership: |
|

SPR
Genomic Map