Assay Details
| Gene Symbol: LOC695267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146698.2 | XP_015002184.2 | 4121 | 86 | ||
| XM_015146699.2 | XP_015002185.2 | 3725 | 86 | |||
| XM_015146702.2 | XP_015002188.2 | 3663 | 86 | |||
| XM_015146704.2 | XP_015002190.2 | 3692 | 86 | |||
| XM_015146705.2 | XP_015002191.2 | 4121 | 86 | |||
| XM_015146706.2 | XP_015002192.2 | 4025 | 86 | |||
| XM_015146707.2 | XP_015002193.2 | 4121 | 86 | |||
| XM_015146708.2 | XP_015002194.2 | 3950 | 86 | |||
| XM_015146709.2 | XP_015002195.2 | 4025 | 86 | |||
| XM_015146710.2 | XP_015002196.2 | 3197 | 86 | |||
| XM_015146711.2 | XP_015002197.2 | 3026 | 86 | |||
| XM_015146713.2 | XP_015002199.2 | 2945 | 86 | |||
| XM_015146714.2 | XP_015002200.2 | 2849 | 86 | |||
| XM_015146715.2 | XP_015002201.2 | 2849 | 86 | |||
| XM_028826127.1 | XP_028681960.1 | 3858 | 86 | |||
| XM_028826128.1 | XP_028681961.1 | 3197 | 86 | |||
| XM_028826130.1 | XP_028681963.1 | 2945 | 86 | |||
More Information
Set Membership: |
|

LOC695267
Genomic Map