Assay Details
Gene Symbol: LOC695267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146698.2 | XP_015002184.2 | 6832 | 153 | ||
XM_015146699.2 | XP_015002185.2 | 6436 | 153 | |||
XM_015146702.2 | XP_015002188.2 | 6374 | 153 | |||
XM_015146704.2 | XP_015002190.2 | 6403 | 153 | |||
XM_015146705.2 | XP_015002191.2 | 6754 | 153 | |||
XM_015146706.2 | XP_015002192.2 | 6736 | 153 | |||
XM_015146707.2 | XP_015002193.2 | 6706 | 153 | |||
XM_015146708.2 | XP_015002194.2 | 6661 | 153 | |||
XM_015146709.2 | XP_015002195.2 | 6610 | 153 | |||
XM_015146710.2 | XP_015002196.2 | 5908 | 153 | |||
XM_015146711.2 | XP_015002197.2 | 5737 | 153 | |||
XM_015146713.2 | XP_015002199.2 | 5656 | 153 | |||
XM_015146714.2 | XP_015002200.2 | 5560 | 153 | |||
XM_015146715.2 | XP_015002201.2 | 5434 | 153 | |||
XM_028826127.1 | XP_028681960.1 | 6569 | 153 | |||
XM_028826128.1 | XP_028681961.1 | 5782 | 153 | |||
XM_028826130.1 | XP_028681963.1 | 5530 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
