Assay Details
| Gene Symbol: TGFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257475.1 | NP_001244404.1 | 1509 | 83 | ||
| XM_015144130.2 | XP_014999616.1 | 2125 | 83 | |||
| XM_028850305.1 | XP_028706138.1 | 433 | 83 | |||
| GenBank mRNA | JU320356.1 | - | 1509 | 83 | ||
| JV044718.1 | - | 1613 | 83 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028850305.1 transcripts.
Set Membership: |
|

TGFB3
Genomic Map