Assay Details
| Gene Symbol: UCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115192.2 | XP_014970678.1 | 1546 | 64 | ||
| XM_015115194.2 | XP_014970680.1 | 1342 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UCP3| Gene Symbol: UCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115192.2 | XP_014970678.1 | 1546 | 64 | ||
| XM_015115194.2 | XP_014970680.1 | 1342 | 64 | |||
Set Membership: |
|