Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146566.2 | XP_015002052.2 | 739 | 84 | ||
| XM_028826032.1 | XP_028681865.1 | 861 | 84 | |||
| XM_028826033.1 | XP_028681866.1 | 869 | 84 | |||
| XM_028826034.1 | XP_028681867.1 | 672 | 84 | |||
| XM_028826035.1 | XP_028681868.1 | 168 | 84 | |||
| GenBank mRNA | JU321694.1 | - | 717 | 84 | ||
| JU471861.1 | - | 670 | 84 | |||
| JV043151.1 | - | 625 | 84 | |||
| JV565014.1 | - | 745 | 84 | |||
| JV704614.1 | - | 747 | 84 | |||
More Information
Set Membership: |
|

STAM
Genomic Map