Assay Details
| Gene Symbol: CDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266610.2 | NP_001253539.1 | 240 | 117 | ||
| XM_015143633.2 | XP_014999119.1 | 270 | 117 | |||
| XM_015143638.2 | XP_014999124.1 | 367 | 117 | |||
| XM_015143642.2 | XP_014999128.1 | 739 | 117 | |||
| XM_028841661.1 | XP_028697494.1 | 228 | 117 | |||
| GenBank mRNA | JU325672.1 | - | 105 | 117 | ||
| JU477412.1 | - | 273 | 117 | |||
| JV048382.1 | - | 240 | 117 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDC7
Genomic Map