Assay Details
| Gene Symbol: PPM1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260836.2 | NP_001247765.1 | 1056 | 71 | ||
| GenBank mRNA | JV048492.1 | - | 1056 | 71 | ||
| JV636423.1 | - | 1036 | 71 | |||
More Information
Set Membership: |
|

PPM1D
Genomic Map