Assay Details
| Gene Symbol: DEGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266006.1 | NP_001252935.1 | 903 | 138 | ||
| XM_028837501.1 | XP_028693334.1 | 1069 | 138 | |||
| XM_028837529.1 | XP_028693362.1 | 860 | 138 | |||
| GenBank mRNA | JU322768.1 | - | 937 | 138 | ||
| JU332241.1 | - | 937 | 138 | |||
| JU332242.1 | - | 937 | 138 | |||
| JU332243.1 | - | 937 | 138 | |||
| JU332244.1 | - | 937 | 138 | |||
| JU332245.1 | - | 937 | 138 | |||
| JU332246.1 | - | 937 | 138 | |||
| JU332247.1 | - | 937 | 138 | |||
| JU332248.1 | - | 937 | 138 | |||
| JU332249.1 | - | 937 | 138 | |||
| JU471089.1 | - | 911 | 138 | |||
| JV045802.1 | - | 903 | 138 | |||
| JV725811.1 | - | 937 | 138 | |||
More Information
Set Membership: |
|

DEGS1
Genomic Map