Assay Details
| Gene Symbol: PDXK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845290.1 | XP_028701123.1 | 934 | 97 | ||
| XM_028845291.1 | XP_028701124.1 | 1056 | 97 | |||
| GenBank mRNA | JU323025.1 | - | 956 | 97 | ||
| JU472309.1 | - | 941 | 97 | |||
| JU472310.1 | - | 941 | 97 | |||
| JV045374.1 | - | 940 | 97 | |||
| JV045375.1 | - | 940 | 97 | |||
More Information
Set Membership: |
|

PDXK
Genomic Map