Assay Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107009.4 | XP_001107009.3 | 508 | 142 | ||
| XM_015110799.2 | XP_014966285.2 | 487 | 142 | |||
| XM_015110800.2 | XP_014966286.1 | 333 | 142 | |||
| GenBank mRNA | JU320689.1 | - | 280 | 142 | ||
| JU472152.1 | - | 433 | 142 | |||
More Information
Set Membership: |
|

KLF7
Genomic Map