Assay Details
| Gene Symbol: VAMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU473789.1 | - | 87 | 124 | ||
| JV045572.1 | - | 97 | 124 | |||
| JV639417.1 | - | 129 | 124 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VAMP8| Gene Symbol: VAMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU473789.1 | - | 87 | 124 | ||
| JV045572.1 | - | 97 | 124 | |||
| JV639417.1 | - | 129 | 124 | |||
Set Membership: |
|