Assay Details
| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852462.1 | XP_028708295.1 | 1212 | 153 | ||
| Gene Symbol: LOC106999707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852463.1 | XP_028708296.1 | 156 | |||
More Information
Set Membership: |
|

TNFRSF10D
Genomic Map