Assay Details
| Gene Symbol: TNFRSF10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144975.2 | XP_015000461.2 | 802 | 148 | ||
| GenBank mRNA | JV664502.1 | - | 337 | 148 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF10A| Gene Symbol: TNFRSF10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144975.2 | XP_015000461.2 | 802 | 148 | ||
| GenBank mRNA | JV664502.1 | - | 337 | 148 | ||
Set Membership: |
|