Assay Details
| Gene Symbol: DPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260828.2 | NP_001247757.1 | 120 | 93 | ||
| GenBank mRNA | JU323319.1 | - | 162 | 93 | ||
| JU477660.1 | - | 122 | 93 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DPM2
Genomic Map