Assay Details
| Gene Symbol: DPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260828.2 | NP_001247757.1 | 221 | 124 | ||
| GenBank mRNA | JU477660.1 | - | 223 | 124 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DPM2
Genomic Map