Assay Details
| Gene Symbol: PROZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087442.4 | XP_001087442.2 | 546 | 107 | ||
| XM_028837371.1 | XP_028693204.1 | 489 | 107 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PROZ| Gene Symbol: PROZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087442.4 | XP_001087442.2 | 546 | 107 | ||
| XM_028837371.1 | XP_028693204.1 | 489 | 107 | |||
Set Membership: |
|