Assay Details
| Gene Symbol: RAB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260953.1 | NP_001247882.1 | 340 | 109 | ||
| XM_015118289.2 | XP_014973775.1 | 424 | 109 | |||
| XM_015118296.2 | XP_014973782.1 | 412 | 109 | |||
| XM_015118304.2 | XP_014973790.1 | 398 | 109 | |||
| XM_015118310.2 | XP_014973796.1 | 166 | 109 | |||
| XM_015118315.2 | XP_014973801.1 | 163 | 109 | |||
| XM_015118321.2 | XP_014973807.1 | 126 | 109 | |||
| GenBank mRNA | JU321140.1 | - | 403 | 109 | ||
| JU472082.1 | - | 340 | 109 | |||
| JV704065.1 | - | 411 | 109 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015118310.2, XM_015118315.2, XM_015118321.2 transcripts.
Set Membership: |
|

RAB29
Genomic Map