Assay Details
| Gene Symbol: ATP6V0E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261126.1 | NP_001248055.1 | 230 | 107 | ||
| GenBank mRNA | JU321368.1 | - | 220 | 107 | ||
| JU473429.1 | - | 230 | 107 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATP6V0E1
Genomic Map